Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCTCCCGCCCCACTCACTTTTG[A/G]TCAAAACCAGCCTCCACAGCCCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608457 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CBX7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CBX7 - chromobox 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175709.3 | 1256 | UTR 3 | NP_783640.1 | |||
XM_005261413.4 | 1256 | UTR 3 | XP_005261470.1 | |||
XM_006724174.3 | 1256 | UTR 3 | XP_006724237.1 | |||
XM_006724175.3 | 1256 | UTR 3 | XP_006724238.1 | |||
XM_006724176.3 | 1256 | UTR 3 | XP_006724239.1 | |||
XM_006724177.3 | 1256 | UTR 3 | XP_006724240.1 | |||
XM_006724178.3 | 1256 | UTR 3 | XP_006724241.1 | |||
XM_011530025.2 | 1256 | UTR 3 | XP_011528327.1 | |||
XM_017028679.1 | 1256 | UTR 3 | XP_016884168.1 |