Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACAGTCTCCCTCCTATTTGGCCA[C/T]TCCAAGCCACAGGCTGCATCAGGGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 190040 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDGFB PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PDGFB - platelet derived growth factor subunit B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002608.3 | 2990 | UTR 3 | NP_002599.1 | |||
NM_033016.3 | 2990 | UTR 3 | NP_148937.1 |
Set Membership: |
HapMap |