Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTAGGCCTCACCTACTCCACTCAA[A/G]CCACCCCATCCCCCTTAGCTTTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608740 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NFAM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NFAM1 - NFAT activating protein with ITAM motif 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318323.1 | 4929 | UTR 3 | NP_001305252.1 | |||
NM_145912.6 | 4929 | UTR 3 | NP_666017.1 | |||
XM_005261374.4 | 4929 | UTR 3 | XP_005261431.1 |