Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGATCCCCATATGCTAGAAAGGGG[C/T]GGCAAGGAAGGTTGTTTCTTGTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603719 MIM: 600647 MIM: 613380 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BUB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BUB3 - BUB3, mitotic checkpoint protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HMX2 - H6 family homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005519.1 | Intron | NP_005510.1 | ||||
XM_005269743.3 | Intron | XP_005269800.1 |
HMX3 - H6 family homeobox 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |