Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTCTGTCTGAGATCTCCTCAGA[A/G]TTCCCTTTACTCTCCACATTTCTGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612869 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATRNL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATRNL1 - attractin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276282.3 | 2145 | Intron | NP_001263211.1 | |||
NM_207303.4 | 2145 | Intron | NP_997186.1 | |||
XM_011539587.1 | 2145 | Intron | XP_011537889.1 | |||
XM_011539588.2 | 2145 | Intron | XP_011537890.1 | |||
XM_011539589.2 | 2145 | Intron | XP_011537891.1 | |||
XM_011539590.2 | 2145 | Intron | XP_011537892.1 | |||
XM_011539591.2 | 2145 | Intron | XP_011537893.1 | |||
XM_017016035.1 | 2145 | Intron | XP_016871524.1 | |||
XM_017016036.1 | 2145 | Intron | XP_016871525.1 | |||
XM_017016037.1 | 2145 | Intron | XP_016871526.1 | |||
XM_017016038.1 | 2145 | UTR 5 | XP_016871527.1 | |||
XM_017016039.1 | 2145 | Intron | XP_016871528.1 | |||
XM_017016040.1 | 2145 | Intron | XP_016871529.1 |
Set Membership: |
HapMap |