Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTCTCCCTGCTTCTTCAATGTG[C/T]CCATTCTAGTGGATCAGTCCCGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611282 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNMBP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DNMBP - dynamin binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318326.1 | Intron | NP_001305255.1 | ||||
NM_001318327.1 | Intron | NP_001305256.1 | ||||
NM_015221.3 | Intron | NP_056036.1 | ||||
XM_006717735.2 | Intron | XP_006717798.1 | ||||
XM_011539559.2 | Intron | XP_011537861.1 |
DNMBP-AS1 - DNMBP antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |