Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAATCACTTCACCTCCCTCACCT[C/T]GGTTTTCTTTATCTCTTACGTGGAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604619 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LGI1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LGI1 - leucine rich glioma inactivated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308275.1 | Intron | NP_001295204.1 | ||||
NM_001308276.1 | Intron | NP_001295205.1 | ||||
NM_005097.3 | Intron | NP_005088.1 | ||||
XM_017016911.1 | Intron | XP_016872400.1 | ||||
XM_017016912.1 | Intron | XP_016872401.1 |
LOC101927032 - uncharacterized LOC101927032 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |