Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTTTATATTTGAGTTTTGAAATT[C/T]AGAATAATCATCAAACTTTTTTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608866 MIM: 600827 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRA10AC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FRA10AC1 - fragile site, folic acid type, rare, fra(10)(q23.3) or fra(10)(q24.2) candidate 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145246.4 | Intron | NP_660289.2 | ||||
XM_005269515.4 | Intron | XP_005269572.1 | ||||
XM_005269516.4 | Intron | XP_005269573.1 | ||||
XM_011539259.2 | Intron | XP_011537561.1 | ||||
XM_011539260.2 | Intron | XP_011537562.1 | ||||
XM_017015656.1 | Intron | XP_016871145.1 | ||||
XM_017015657.1 | Intron | XP_016871146.1 | ||||
XM_017015658.1 | Intron | XP_016871147.1 | ||||
XM_017015659.1 | Intron | XP_016871148.1 | ||||
XM_017015660.1 | Intron | XP_016871149.1 | ||||
XM_017015661.1 | Intron | XP_016871150.1 | ||||
XM_017015662.1 | Intron | XP_016871151.1 |
PDE6C - phosphodiesterase 6C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |