Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGGAAAGAAAGAAGTTAAAGACA[C/T]ACATAATATACTTTAATCCTAGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608866 MIM: 600827 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FRA10AC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FRA10AC1 - fragile site, folic acid type, rare, fra(10)(q23.3) or fra(10)(q24.2) candidate 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145246.4 | 1796 | UTR 3 | NP_660289.2 | |||
XM_005269515.4 | 1796 | Intron | XP_005269572.1 | |||
XM_005269516.4 | 1796 | Intron | XP_005269573.1 | |||
XM_011539259.2 | 1796 | Intron | XP_011537561.1 | |||
XM_011539260.2 | 1796 | Intron | XP_011537562.1 | |||
XM_017015656.1 | 1796 | UTR 3 | XP_016871145.1 | |||
XM_017015657.1 | 1796 | UTR 3 | XP_016871146.1 | |||
XM_017015658.1 | 1796 | UTR 3 | XP_016871147.1 | |||
XM_017015659.1 | 1796 | UTR 3 | XP_016871148.1 | |||
XM_017015660.1 | 1796 | UTR 3 | XP_016871149.1 | |||
XM_017015661.1 | 1796 | Intron | XP_016871150.1 | |||
XM_017015662.1 | 1796 | Intron | XP_016871151.1 |
PDE6C - phosphodiesterase 6C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |