Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCTTTCACAATCATTCATTCATT[A/G]GATAAGCATTTCTTGAGTATCTACT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609672 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXOC6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EXOC6 - exocyst complex component 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013848.3 | Intron | NP_001013870.1 | ||||
NM_001319194.1 | Intron | NP_001306123.1 | ||||
NM_001319195.1 | Intron | NP_001306124.1 | ||||
NM_001319200.1 | Intron | NP_001306129.1 | ||||
NM_019053.5 | Intron | NP_061926.3 | ||||
XM_017016344.1 | Intron | XP_016871833.1 | ||||
XM_017016345.1 | Intron | XP_016871834.1 | ||||
XM_017016346.1 | Intron | XP_016871835.1 | ||||
XM_017016347.1 | Intron | XP_016871836.1 | ||||
XM_017016348.1 | Intron | XP_016871837.1 |
Set Membership: |
HapMap |