Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATACCTTATGTCTCCTTGGAGCAT[C/T]TACTGCCAACATTTCCTAGGCACAA
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 146680 | |||||||||||||||||||||||||||||
Literature Links: |
IDE PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
IDE - insulin degrading enzyme | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165946.1 | 1499 | Missense Mutation | AAT,GAT | N,D 392 | NP_001159418.1 | |
NM_001322793.1 | 1499 | Missense Mutation | AAT,GAT | N,D 947 | NP_001309722.1 | |
NM_001322794.1 | 1499 | Missense Mutation | AAT,GAT | N,D 908 | NP_001309723.1 | |
NM_001322795.1 | 1499 | Missense Mutation | AAT,GAT | N,D 906 | NP_001309724.1 | |
NM_001322796.1 | 1499 | Missense Mutation | AAT,GAT | N,D 906 | NP_001309725.1 | |
NM_001322797.1 | 1499 | Missense Mutation | AAT,GAT | N,D 392 | NP_001309726.1 | |
NM_004969.3 | 1499 | Missense Mutation | AAT,GAT | N,D 947 | NP_004960.2 | |
XM_017016187.1 | 1499 | Missense Mutation | AAT,GAT | N,D 906 | XP_016871676.1 | |
XM_017016188.1 | 1499 | Missense Mutation | AAT,GAT | N,D 906 | XP_016871677.1 | |
XM_017016189.1 | 1499 | Missense Mutation | AAT,GAT | N,D 906 | XP_016871678.1 | |
XM_017016190.1 | 1499 | Missense Mutation | AAT,GAT | N,D 906 | XP_016871679.1 |
Set Membership: |
HapMap |