Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610606 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPEB3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CPEB3 - cytoplasmic polyadenylation element binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178137.1 | Intron | NP_001171608.1 | ||||
NM_014912.4 | Intron | NP_055727.3 | ||||
XM_005269630.4 | Intron | XP_005269687.1 | ||||
XM_006717714.3 | Intron | XP_006717777.1 | ||||
XM_006717715.3 | Intron | XP_006717778.1 | ||||
XM_011539514.2 | Intron | XP_011537816.1 | ||||
XM_011539515.2 | Intron | XP_011537817.1 | ||||
XM_011539517.2 | Intron | XP_011537819.1 | ||||
XM_011539518.2 | Intron | XP_011537820.1 | ||||
XM_011539519.2 | Intron | XP_011537821.1 | ||||
XM_011539520.2 | Intron | XP_011537822.1 | ||||
XM_011539522.2 | Intron | XP_011537824.1 | ||||
XM_011539523.2 | Intron | XP_011537825.1 | ||||
XM_017015927.1 | Intron | XP_016871416.1 |
Set Membership: |
HapMap |