Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGGGGAAGTTTCTTGGGCTTAC[A/G]TGTTTTTAAAAAAGGCCAGACTTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604492 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C5orf15 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C5orf15 - chromosome 5 open reading frame 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105379183 - uncharacterized LOC105379183 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VDAC1 - voltage dependent anion channel 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003374.2 | Intron | NP_003365.1 | ||||
XM_005272075.3 | Intron | XP_005272132.1 | ||||
XM_017009821.1 | Intron | XP_016865310.1 | ||||
XM_017009822.1 | Intron | XP_016865311.1 | ||||
XM_017009823.1 | Intron | XP_016865312.1 |
Set Membership: |
HapMap |