Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACAAGCTCCCCATTGATGCCAAT[G/T]CTGTTGGTCCATAGATCTCGACAAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151690 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANXA1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ANXA1 - annexin A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000700.2 | Intron | NP_000691.1 | ||||
XM_011518609.1 | Intron | XP_011516911.1 | ||||
XM_017014657.1 | Intron | XP_016870146.1 |
Set Membership: |
HapMap |