Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAACAAACCAGATACAAAAGAGCA[C/T]ACTATGTGATTCCATCTATAGAGAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABHD17B PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ABHD17B - abhydrolase domain containing 17B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C9orf85 - chromosome 9 open reading frame 85 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182505.3 | Intron | NP_872311.2 | ||||
XM_011518226.2 | Intron | XP_011516528.1 |