Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGGGGGATTTGTTCCCAACATCC[A/G]TCTCTACTAAAGATACAAAAGCTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600391 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCNT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GCNT1 - glucosaminyl (N-acetyl) transferase 1, core 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001097633.1 | Intron | NP_001091102.1 | ||||
NM_001097634.1 | Intron | NP_001091103.1 | ||||
NM_001097635.1 | Intron | NP_001091104.1 | ||||
NM_001097636.1 | Intron | NP_001091105.1 | ||||
NM_001490.4 | Intron | NP_001481.2 | ||||
XM_017014620.1 | Intron | XP_016870109.1 | ||||
XM_017014621.1 | Intron | XP_016870110.1 |
Set Membership: |
HapMap |