Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGACACACTGGGACCTTCTTTCCT[G/T]TTCCAAATGGCTTCAGCCTTGCAGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600831 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DAPK1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DAPK1 - death associated protein kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288729.1 | Intron | NP_001275658.1 | ||||
NM_001288730.1 | Intron | NP_001275659.1 | ||||
NM_001288731.1 | Intron | NP_001275660.1 | ||||
NM_004938.3 | Intron | NP_004929.2 | ||||
XM_005251757.3 | Intron | XP_005251814.1 | ||||
XM_017014360.1 | Intron | XP_016869849.1 |
Set Membership: |
HapMap |