Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAAAAGAAATACTAAAGGGTGTTC[C/T]TCAGTAAGAAGCAAAATAATTCTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD19P PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANKRD19P - ankyrin repeat domain 19, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF484 - zinc finger protein 484 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007101.2 | Intron | NP_001007102.1 | ||||
NM_001261458.1 | Intron | NP_001248387.1 | ||||
NM_001261459.1 | Intron | NP_001248388.1 | ||||
NM_001261460.1 | Intron | NP_001248389.1 | ||||
NM_031486.2 | Intron | NP_113674.1 | ||||
XM_006717299.3 | Intron | XP_006717362.1 | ||||
XM_011519073.2 | Intron | XP_011517375.1 | ||||
XM_011519074.2 | Intron | XP_011517376.1 | ||||
XM_011519075.2 | Intron | XP_011517377.1 | ||||
XM_017015178.1 | Intron | XP_016870667.1 | ||||
XM_017015179.1 | Intron | XP_016870668.1 | ||||
XM_017015180.1 | Intron | XP_016870669.1 | ||||
XM_017015181.1 | Intron | XP_016870670.1 |
Set Membership: |
HapMap |