Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAATGTGTTTATTAACCAACACAT[G/T]TATTTACATGAGCAAAATATTTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614790 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
WTIP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
WTIP - Wilms tumor 1 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080436.1 | Intron | NP_001073905.1 | ||||
XM_006723014.3 | Intron | XP_006723077.1 | ||||
XM_011526452.2 | Intron | XP_011524754.1 | ||||
XM_011526453.2 | Intron | XP_011524755.1 |
Set Membership: |
HapMap |