Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAACATGTTGGAAGATCAAAGGC[A/G]CAGGTAACTATATTTCTCATACCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615661 MIM: 613295 | ||||||||||||||||||||
Literature Links: |
PDCD2L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PDCD2L - programmed cell death 2 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBA2 - ubiquitin like modifier activating enzyme 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005499.2 | 289 | Silent Mutation | GCA,GCG | A,A 73 | NP_005490.1 | |
XM_005258404.3 | 289 | Silent Mutation | GCA,GCG | A,A 185 | XP_005258461.2 | |
XM_006722962.2 | 289 | Intron | XP_006723025.1 | |||
XM_011526304.2 | 289 | Silent Mutation | GCA,GCG | A,A 185 | XP_011524606.1 | |
XM_017026134.1 | 289 | Silent Mutation | GCA,GCG | A,A 185 | XP_016881623.1 |