Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTCCTTAGGTGAGGGGCATGAAT[C/T]TGGGTTCCCACGAGTCTCCCTCTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615601 | ||||||||||||||||||||
Literature Links: |
GADL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GADL1 - glutamate decarboxylase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207359.2 | Intron | NP_997242.2 | ||||
XM_017006297.1 | Intron | XP_016861786.1 | ||||
XM_017006298.1 | Intron | XP_016861787.1 |