Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCAGGAGCCGCCTTTCCGATTCC[C/T]TACGATGCGGGTGCTGAGCTATGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612811 | ||||||||||||||||||||
Literature Links: |
LRFN5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRFN5 - leucine rich repeat and fibronectin type III domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152447.3 | 1729 | UTR 5 | NP_689660.2 | |||
XM_005267369.3 | 1729 | UTR 5 | XP_005267426.1 | |||
XM_017021048.1 | 1729 | Intron | XP_016876537.1 | |||
XM_017021049.1 | 1729 | UTR 5 | XP_016876538.1 | |||
XM_017021050.1 | 1729 | UTR 5 | XP_016876539.1 |