Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCTGGACATGCACTTTCTTAGG[A/G]GGCTTTGTGTTGCTGGTGGTGTTCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613317 MIM: 612028 MIM: 600654 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCAF11 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DCAF11 - DDB1 and CUL4 associated factor 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EMC9 - ER membrane protein complex subunit 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FITM1 - fat storage inducing transmembrane protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_203402.2 | 410 | Silent Mutation | GGA,GGG | G,G 104 | NP_981947.1 |
PSME1 - proteasome activator subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |