Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCGGCCAGGCGGGCGCGCTCGGC[A/C]TCCAGCCCGCGCCGCTGCTGCAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605688 MIM: 162080 | ||||||||||||||||||||
Literature Links: |
CPNE6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CPNE6 - copine 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NRL - neural retina leucine zipper | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006177.3 | 479 | Missense Mutation | GAG,GAT | E,D 188 | NP_006168.1 | |
XM_005267708.4 | 479 | Missense Mutation | GAG,GAT | E,D 188 | XP_005267765.1 | |
XM_005267709.3 | 479 | Missense Mutation | GAG,GAT | E,D 188 | XP_005267766.1 | |
XM_005267710.3 | 479 | Missense Mutation | GAG,GAT | E,D 188 | XP_005267767.1 | |
XM_011536801.2 | 479 | Missense Mutation | GAG,GAT | E,D 290 | XP_011535103.2 | |
XM_011536802.1 | 479 | Missense Mutation | GAG,GAT | E,D 188 | XP_011535104.1 | |
XM_011536804.2 | 479 | Missense Mutation | GAG,GAT | E,D 188 | XP_011535106.1 | |
XM_011536805.2 | 479 | Missense Mutation | GAG,GAT | E,D 188 | XP_011535107.1 | |
XM_011536806.2 | 479 | Missense Mutation | GAG,GAT | E,D 185 | XP_011535108.2 | |
XM_017021350.1 | 479 | Missense Mutation | GAG,GAT | E,D 83 | XP_016876839.1 |