Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGAGGTCAGCTGGGCTGAGTTCA[A/G]GTGTCACTGAATGGAACTCAGTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601066 MIM: 603593 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OXA1L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OXA1L - OXA1L, mitochondrial inner membrane protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC7A7 - solute carrier family 7 member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126105.2 | Intron | NP_001119577.1 | ||||
NM_001126106.2 | Intron | NP_001119578.1 | ||||
XM_006720302.1 | Intron | XP_006720365.1 | ||||
XM_011537298.2 | Intron | XP_011535600.1 | ||||
XM_011537299.1 | Intron | XP_011535601.1 |
Set Membership: |
HapMap |