Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAACAACAACAACAAAACCCTAG[G/T]AGAGCAAAAGGAGGTGGAGAGAGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611612 | ||||||||||||||||||||
Literature Links: |
THTPA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
THTPA - thiamine triphosphatase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126339.3 | Intron | NP_001119811.1 | ||||
NM_001256062.2 | Intron | NP_001242991.1 | ||||
NM_001256321.2 | Intron | NP_001243250.1 | ||||
NM_001256322.2 | Intron | NP_001243251.1 | ||||
NM_001256323.2 | Intron | NP_001243252.1 | ||||
NM_024328.5 | Intron | NP_077304.1 |
ZFHX2 - zinc finger homeobox 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |