Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTCATGCTAAAATAACCGTATCA[G/T]TCAAATTCAGCACAAATCTCATGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607219 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNTN5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CNTN5 - contactin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243270.1 | Intron | NP_001230199.1 | ||||
NM_001243271.1 | Intron | NP_001230200.1 | ||||
NM_014361.3 | Intron | NP_055176.1 | ||||
NM_175566.2 | Intron | NP_780775.1 | ||||
XM_011542871.1 | Intron | XP_011541173.1 | ||||
XM_011542873.1 | Intron | XP_011541175.1 | ||||
XM_017017926.1 | Intron | XP_016873415.1 | ||||
XM_017017927.1 | Intron | XP_016873416.1 | ||||
XM_017017928.1 | Intron | XP_016873417.1 | ||||
XM_017017929.1 | Intron | XP_016873418.1 |