Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTACCCTTCAACCCTCCCATTT[C/T]ATCCTAGCACCCACCTTGCCTCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607666 MIM: 614634 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANGPTL5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANGPTL5 - angiopoietin like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178127.4 | Intron | NP_835228.2 |
CEP126 - centrosomal protein 126 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020802.3 | Intron | NP_065853.3 | ||||
XM_005271627.4 | Intron | XP_005271684.2 |
Set Membership: |
HapMap |