Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCTGCCACTGCTCTATTACCTG[G/T]TCACTTCTGTCCCTCATTTCTCCTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179410 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RDX PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RDX - radixin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001260492.1 | Intron | NP_001247421.1 | ||||
NM_001260493.1 | Intron | NP_001247422.1 | ||||
NM_001260494.1 | Intron | NP_001247423.1 | ||||
NM_001260495.1 | Intron | NP_001247424.1 | ||||
NM_001260496.1 | Intron | NP_001247425.1 | ||||
NM_002906.3 | Intron | NP_002897.1 |
Set Membership: |
HapMap |