Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTGGCTCTCATTGCGTTCCATT[C/T]TGAAAGCTTCGTGTCTCTGCATTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605673 | ||||||||||||||||||||
Literature Links: |
BTG4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BTG4 - BTG anti-proliferation factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017589.3 | Intron | NP_060059.1 | ||||
XM_011542876.2 | Intron | XP_011541178.1 | ||||
XM_011542878.2 | Intron | XP_011541180.1 | ||||
XM_011542879.2 | Intron | XP_011541181.1 | ||||
XM_011542880.2 | Intron | XP_011541182.1 |