Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACCTTACTGAATAACCCTTTAAT[A/G]TACTGTTAGATCAATTGCAGGACCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615001 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZC3H12C PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZC3H12C - zinc finger CCCH-type containing 12C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033390.1 | Intron | NP_203748.1 | ||||
XM_005271715.3 | Intron | XP_005271772.1 | ||||
XM_011543055.2 | Intron | XP_011541357.1 | ||||
XM_017018477.1 | Intron | XP_016873966.1 |
Set Membership: |
HapMap |