Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGCTTCCCTCTTCACGAACTCT[C/T]AAAGAAAAGGAAGGATAAAACCTAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 188230 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
THY1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
THY1 - Thy-1 cell surface antigen | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001311160.1 | 1801 | UTR 3 | NP_001298089.1 | |||
NM_001311162.1 | 1801 | UTR 3 | NP_001298091.1 | |||
NM_006288.4 | 1801 | UTR 3 | NP_006279.2 |
USP2-AS1 - USP2 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |