Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATTTCAATTTTGAGCCTATAGGA[C/T]ATCTCATACTGCAAAGGAAAAGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604763 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGEF12 - Rho guanine nucleotide exchange factor 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198665.1 | Intron | NP_001185594.1 | ||||
NM_001301084.1 | Intron | NP_001288013.1 | ||||
NM_015313.2 | Intron | NP_056128.1 | ||||
XM_006718805.3 | Intron | XP_006718868.1 | ||||
XM_011542720.2 | Intron | XP_011541022.1 | ||||
XM_017017420.1 | Intron | XP_016872909.1 | ||||
XM_017017421.1 | Intron | XP_016872910.1 | ||||
XM_017017422.1 | Intron | XP_016872911.1 |