Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTTAGAAAGCTGTACACTTTTT[A/T]AAAAAATTTGCACTTATAAATAATA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606871 MIM: 609276 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
JAM3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
JAM3 - junctional adhesion molecule 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205329.1 | 5554 | Intron | NP_001192258.1 | |||
NM_032801.4 | 5554 | Intron | NP_116190.3 |
NCAPD3 - non-SMC condensin II complex subunit D3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015261.2 | 5554 | UTR 3 | NP_056076.1 | |||
XM_011542719.2 | 5554 | UTR 3 | XP_011541021.1 |
Set Membership: |
HapMap |