Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAACTTACAACATACAACAAGCTA[C/T]GTTGTTAAGCTCATAGAAGGCCATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLB1L3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLB1L3 - galactosidase beta 1 like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080407.2 | Intron | NP_001073876.2 | ||||
XM_011542566.2 | Intron | XP_011540868.1 | ||||
XM_011542567.2 | Intron | XP_011540869.1 | ||||
XM_011542568.2 | Intron | XP_011540870.1 | ||||
XM_011542569.2 | Intron | XP_011540871.1 | ||||
XM_011542570.2 | Intron | XP_011540872.1 | ||||
XM_011542571.2 | Intron | XP_011540873.1 | ||||
XM_011542572.1 | Intron | XP_011540874.1 | ||||
XM_011542573.1 | Intron | XP_011540875.1 | ||||
XM_011542574.2 | Intron | XP_011540876.1 | ||||
XM_017017155.1 | Intron | XP_016872644.1 | ||||
XM_017017156.1 | Intron | XP_016872645.1 |