Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTGGAAGAGATGAGAAAAATCA[A/C]CCTAGTGAAGCAGGGGGCGGAGGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613773 | ||||||||||||||||||||
Literature Links: |
IGSF9B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IGSF9B - immunoglobulin superfamily member 9B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277285.1 | 3106 | UTR 3 | NP_001264214.1 | |||
XM_006718795.3 | 3106 | UTR 3 | XP_006718858.1 | |||
XM_011542691.2 | 3106 | UTR 3 | XP_011540993.1 | |||
XM_011542692.2 | 3106 | UTR 3 | XP_011540994.1 | |||
XM_011542694.2 | 3106 | Intron | XP_011540996.1 | |||
XM_011542695.2 | 3106 | UTR 3 | XP_011540997.1 | |||
XM_017017393.1 | 3106 | UTR 3 | XP_016872882.1 | |||
XM_017017394.1 | 3106 | Intron | XP_016872883.1 | |||
XM_017017395.1 | 3106 | Intron | XP_016872884.1 | |||
XM_017017396.1 | 3106 | UTR 3 | XP_016872885.1 |
MIR4697HG - MIR4697 host gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |