Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTGCTGATAAACTTCTTAAAAGA[A/C]TTCTTGATCTCTTACCTAATTTTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608062 MIM: 611072 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCDC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCDC1 - doublecortin domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DNAJC24 - DnaJ heat shock protein family (Hsp40) member C24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181706.4 | Intron | NP_859057.4 | ||||
XM_017017216.1 | Intron | XP_016872705.1 |