Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATCAGCTGATATAATCAAGTGAG[A/G]ATGTGGGGAGAAATTGTAGGTTCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603433 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC644656 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC644656 - uncharacterized LOC644656 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF143 - zinc finger protein 143 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282656.1 | Intron | NP_001269585.1 | ||||
NM_001282657.1 | Intron | NP_001269586.1 | ||||
NM_003442.5 | Intron | NP_003433.3 | ||||
XM_005253121.1 | Intron | XP_005253178.1 | ||||
XM_005253122.1 | Intron | XP_005253179.1 | ||||
XM_005253127.1 | Intron | XP_005253184.1 | ||||
XM_011520349.2 | Intron | XP_011518651.1 | ||||
XM_017018252.1 | Intron | XP_016873741.1 | ||||
XM_017018253.1 | Intron | XP_016873742.1 | ||||
XM_017018254.1 | Intron | XP_016873743.1 | ||||
XM_017018255.1 | Intron | XP_016873744.1 | ||||
XM_017018256.1 | Intron | XP_016873745.1 |
Set Membership: |
HapMap |