Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTAAAGCCAACTGATTAGGGTAT[A/T]CTGGTATATCTCCGCAACAACTTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR52N4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR52N4 - olfactory receptor family 52 subfamily N member 4 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005175.3 | 879 | Intron | NP_001005175.3 | |||
XM_017017711.1 | 879 | UTR 5 | XP_016873200.1 | |||
XM_017017712.1 | 879 | UTR 5 | XP_016873201.1 | |||
XM_017017713.1 | 879 | UTR 5 | XP_016873202.1 |