Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTGCTGGTTCCACACCCTTCCCA[A/G]CCGGGACCCAAGGGTCCTCTCCAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602565 MIM: 608529 | ||||||||||||||||||||
Literature Links: |
CCL25 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCL25 - C-C motif chemokine ligand 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201359.1 | Intron | NP_001188288.1 | ||||
NM_005624.3 | Intron | NP_005615.2 | ||||
XM_011528177.2 | Intron | XP_011526479.1 | ||||
XM_017027119.1 | Intron | XP_016882608.1 | ||||
XM_017027120.1 | Intron | XP_016882609.1 | ||||
XM_017027121.1 | Intron | XP_016882610.1 | ||||
XM_017027122.1 | Intron | XP_016882611.1 |
FBN3 - fibrillin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |