Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACGTCCCCGAGACCAGGGCACGG[C/T]CCGCGAGGCTGCTCTTGAGATGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601825 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MUM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MUM1 - melanoma associated antigen (mutated) 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFS7 - NADH:ubiquinone oxidoreductase core subunit S7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024407.4 | Intron | NP_077718.3 | ||||
XM_005259556.4 | Intron | XP_005259613.2 | ||||
XM_017026768.1 | Intron | XP_016882257.1 |
Set Membership: |
HapMap |