Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTAATAATTAATTTCTGAAATCT[A/G]GTCATATTCTTGTGCTTGAAACCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC16A14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC16A14 - solute carrier family 16 member 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152527.4 | 2297 | UTR 3 | NP_689740.2 | |||
XM_005246353.3 | 2297 | UTR 3 | XP_005246410.1 | |||
XM_011510751.2 | 2297 | UTR 3 | XP_011509053.1 | |||
XM_011510752.1 | 2297 | Intron | XP_011509054.1 | |||
XM_017003480.1 | 2297 | UTR 3 | XP_016858969.1 |
Set Membership: |
HapMap |