Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCCACTCCATTGAGAAGACAGAA[A/G]CCACAGGCCAGAAACACTCCGCTCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114780 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CA6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CA6 - carbonic anhydrase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001215.3 | Intron | NP_001206.2 | ||||
NM_001270500.1 | Intron | NP_001257429.1 | ||||
NM_001270501.1 | Intron | NP_001257430.1 | ||||
NM_001270502.1 | Intron | NP_001257431.1 | ||||
XM_011542083.2 | Intron | XP_011540385.1 | ||||
XM_011542084.2 | Intron | XP_011540386.1 |