Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCTCCTTGCAGCATCCGTGTGAG[C/T]TGTGCGTGAATAAAAGAAATCGTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 154100 MIM: 616695 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MDH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MDH2 - malate dehydrogenase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282403.1 | 86 | Intron | NP_001269332.1 | |||
NM_001282404.1 | 86 | Intron | NP_001269333.1 | |||
NM_005918.3 | 86 | Intron | NP_005909.2 |
STYXL1 - serine/threonine/tyrosine interacting-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317785.1 | 86 | Intron | NP_001304714.1 | |||
NM_001317786.1 | 86 | Intron | NP_001304715.1 | |||
NM_001317787.1 | 86 | Intron | NP_001304716.1 | |||
NM_001317788.1 | 86 | Intron | NP_001304717.1 | |||
NM_001317789.1 | 86 | Intron | NP_001304718.1 | |||
NM_016086.2 | 86 | Intron | NP_057170.1 | |||
XM_011516292.2 | 86 | Intron | XP_011514594.1 | |||
XM_011516293.2 | 86 | Intron | XP_011514595.1 | |||
XM_011516300.1 | 86 | Intron | XP_011514602.1 | |||
XM_017012296.1 | 86 | Intron | XP_016867785.1 | |||
XM_017012297.1 | 86 | Intron | XP_016867786.1 | |||
XM_017012298.1 | 86 | UTR 5 | XP_016867787.1 | |||
XM_017012299.1 | 86 | Intron | XP_016867788.1 | |||
XM_017012300.1 | 86 | Intron | XP_016867789.1 | |||
XM_017012301.1 | 86 | Intron | XP_016867790.1 | |||
XM_017012302.1 | 86 | Intron | XP_016867791.1 |