Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTATTTATTTCTACAGTCTCTC[A/G]TCTCCGCATAGAGAGAGCAAAACCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF600 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF600 - zinc finger protein 600 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321866.1 | Intron | NP_001308795.1 | ||||
NM_001321867.1 | Intron | NP_001308796.1 | ||||
NM_198457.3 | Intron | NP_940859.3 | ||||
XM_017026390.1 | Intron | XP_016881879.1 | ||||
XM_017026391.1 | Intron | XP_016881880.1 | ||||
XM_017026392.1 | Intron | XP_016881881.1 |
Set Membership: |
HapMap |