Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAGAGTAGAAATTAAGATACTAT[A/G]CTTTCTTTTTCAATAAAAAGCTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606498 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MS4A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MS4A3 - membrane spanning 4-domains A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031666.1 | Intron | NP_001026836.1 | ||||
NM_001031809.1 | Intron | NP_001026979.1 | ||||
NM_006138.4 | Intron | NP_006129.4 | ||||
XM_011545363.2 | Intron | XP_011543665.1 |
Set Membership: |
HapMap |