Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTGGAAAGGACGCTCTATTTCC[C/T]TCGTGTCTCTACCTGGTAGGTAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613277 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM216 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TMEM216 - transmembrane protein 216 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001173990.2 | 503 | Intron | NP_001167461.1 | |||
NM_001173991.2 | 503 | Intron | NP_001167462.1 | |||
NM_016499.5 | 503 | Intron | NP_057583.2 | |||
XM_005274039.3 | 503 | UTR 5 | XP_005274096.1 | |||
XM_017017882.1 | 503 | UTR 5 | XP_016873371.1 |