Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCTGTTCATTTATTCATGGGATA[A/T]ATCAGCTTTGAGTCCTGTGTGCTTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612163 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TPCN2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TPCN2 - two pore segment channel 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139075.3 | Intron | NP_620714.2 | ||||
XM_005273824.3 | Intron | XP_005273881.1 | ||||
XM_005273826.3 | Intron | XP_005273883.1 | ||||
XM_005273830.3 | Intron | XP_005273887.1 | ||||
XM_005273831.3 | Intron | XP_005273888.1 | ||||
XM_005273832.3 | Intron | XP_005273889.1 | ||||
XM_011544802.2 | Intron | XP_011543104.1 | ||||
XM_011544805.1 | Intron | XP_011543107.1 | ||||
XM_011544806.1 | Intron | XP_011543108.1 | ||||
XM_011544807.2 | Intron | XP_011543109.1 | ||||
XM_011544808.2 | Intron | XP_011543110.1 | ||||
XM_017017328.1 | Intron | XP_016872817.1 | ||||
XM_017017329.1 | Intron | XP_016872818.1 | ||||
XM_017017330.1 | Intron | XP_016872819.1 | ||||
XM_017017331.1 | Intron | XP_016872820.1 | ||||
XM_017017332.1 | Intron | XP_016872821.1 | ||||
XM_017017333.1 | Intron | XP_016872822.1 | ||||
XM_017017334.1 | Intron | XP_016872823.1 | ||||
XM_017017335.1 | Intron | XP_016872824.1 | ||||
XM_017017336.1 | Intron | XP_016872825.1 |
Set Membership: |
HapMap |