Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGGATCAGACGTGGGCGATCAGA[C/T]GTGGGCAATATCTTAGATTTGCCAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 136425 MIM: 600829 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOLR2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FOLR2 - folate receptor beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
INPPL1 - inositol polyphosphate phosphatase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001567.3 | Intron | NP_001558.3 | ||||
XM_005273978.3 | Intron | XP_005274035.1 | ||||
XM_005273979.4 | Intron | XP_005274036.1 | ||||
XM_011544999.2 | Intron | XP_011543301.1 | ||||
XM_011545000.2 | Intron | XP_011543302.1 | ||||
XM_017017673.1 | Intron | XP_016873162.1 |