Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACTTGCTGCTGAGCTAAGCTAGA[A/G]CACTATGTAAGCTGGCTGGAAGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602451 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
P2RY6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
P2RY6 - pyrimidinergic receptor P2Y6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277204.1 | Intron | NP_001264133.1 | ||||
NM_001277205.1 | Intron | NP_001264134.1 | ||||
NM_001277206.1 | Intron | NP_001264135.1 | ||||
NM_001277207.1 | Intron | NP_001264136.1 | ||||
NM_001277208.1 | Intron | NP_001264137.1 | ||||
NM_176796.2 | Intron | NP_789766.1 | ||||
NM_176797.2 | Intron | NP_789767.1 | ||||
NM_176798.2 | Intron | NP_789768.1 | ||||
XM_005274022.3 | Intron | XP_005274079.1 | ||||
XM_006718571.3 | Intron | XP_006718634.1 | ||||
XM_011545076.2 | Intron | XP_011543378.1 | ||||
XM_011545077.2 | Intron | XP_011543379.1 | ||||
XM_011545079.2 | Intron | XP_011543381.1 |
Set Membership: |
HapMap |